Thermo Scientific™ Nalgene™ Pipette Washers
Offers wide range of cycling speeds, operating with water input from 1.5 to 12 liters per minute
  • Water inlet at the top eliminates back-siphoning to prevent contamination
  • Durable HDPE will provide years of use in your lab
HDPE washers include top inlet to prevent back-siphoning of contaminated water. Connect directly to drain line. Use water flow as low as 1.5 liters/minute.
Washers include 18” L x 1” ID inlet and 4’ x ½” ID PVC tubing.
108,000.00 - ₹129,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Compatible with Wetted Matterial
Availability Pricing
Thermo Scientific Nalgene Pipette Washer for Up To 16" L Pipets WW-06185-00 Mfr # 5245-0040
COMPARE
30 1/2 Pipets up to 16\" L HDPE and PVC
₹108,000.00INR / Each
Thermo Scientific Nalgene Pipette Washer for Up To 24" L Pipets WW-06190-00 Mfr # 5245-0050
COMPARE
38 Pipets up to 24\" L HDPE and PVC
₹129,000.00INR / Each