Thermo Scientific™ Nalgene™ Griffin Low-Form PFA Beaker
Highly resistant to most chemicals
  • No-drip single-spout design for precision pouring control
  • Autoclavable
These Thermo Scientific™ Nalgene™ beakers are constructed of PFA, which offers superior chemical resistance to concentrated acids and strong oxidizing agents. Molded from chemically resistant perfluoroalkoxy resin and featuring a heavy-wall design, these beakers are ideal for continual lab usage.
Beakers can withstand temperatures from -454 to 482°F (-270 to 250°C).
38,000.00 - ₹97,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Overall Height (in)
Availability Pricing
Thermo Scientific Nalgene 1510-0250 Griffin low-form beaker of PFA, 250 mL WW-06002-55 Mfr # 1510-0250
COMPARE
250 Perfluoroalkoxy (PFA) 3 7/8
₹38,000.00INR / Each
Thermo Scientific Nalgene 1510-1000 Griffin low-form beaker of PFA, 1000 mL WW-06002-85 Mfr # 1510-1000
COMPARE
1000 Perfluoroalkoxy (PFA) 6
₹97,300.00INR / Each