Thermo Scientific™ Nalgene™ Griffin Low-Form Polypropylene Beakers
Durable construction withstands temperatures to 275°F (135°C)
  • Ribs for easy stacking
  • Silk-screened with graduations, size code, resin code, catalog number, max. temperature and “No Flame” symbol for easy identification and safe use
  • Autoclavable
  • Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy
Carry out precise and efficient pouring with Thermo Scientific™ Nalgene™ polypropylene (PP) griffin low-form beakers, featuring a single-spout design and easy-to-read silk-screened graduations.
9,300.00 - ₹15,900.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Thermo Scientific Nalgene 1201-0030 polypropylene Griffin low-form beaker, 30 mL WW-06000-20 Mfr # 1201-0030
COMPARE
30 5 Polypropylene(PP)
₹9,900.00INR / Pkg of 12
Thermo Scientific Nalgene 1201-0050 polypropylene Griffin low-form beaker, 50 mL WW-06000-25 Mfr # 1201-0050
COMPARE
50 5 Polypropylene(PP)
₹9,300.00INR / Pkg of 12
Thermo Scientific Nalgene 1201-0100 polypropylene Griffin low-form beaker, 100 mL WW-06000-30 Mfr # 1201-0100
COMPARE
100 10 Polypropylene(PP)
₹12,100.00INR / Pkg of 12
Thermo Scientific Nalgene 1201-0150 polypropylene Griffin low-form beaker, 150 mL WW-06000-35 Mfr # 1201-0150
COMPARE
150 10 Polypropylene(PP)
₹15,600.00INR / Pkg of 12
Thermo Scientific Nalgene 1201-0250 polypropylene Griffin low-form beaker, 250 mL WW-06000-40 Mfr # 1201-0250
COMPARE
250 25 Polypropylene(PP)
₹10,800.00INR / Pkg of 6
Thermo Scientific Nalgene 1201-0400 polypropylene Griffin low-form beaker, 400 mL, 6/pk WW-06000-50 Mfr # 1201-0400
COMPARE
400 25 Polypropylene(PP)
₹15,600.00INR / Pkg of 6
Thermo Scientific Nalgene 1201-0600 polypropylene Griffin low-form beaker, 600 mL WW-06000-55 Mfr # 1201-0600
COMPARE
600 50 Polypropylene(PP)
₹13,400.00INR / Pkg of 4
Thermo Scientific Nalgene 1201-1000 polypropylene Griffin low-form beaker, 1000 mL WW-06000-60 Mfr # 1201-1000
COMPARE
1000 50 Polypropylene(PP)
₹12,800.00INR / Pkg of 3
Thermo Scientific Nalgene 1201-2000 polypropylene Griffin low-form beaker, 2000 mL WW-06000-70 Mfr # 1201-2000
COMPARE
2000 100 Polypropylene(PP)
₹9,300.00INR / Each
Thermo Scientific Nalgene 1201-4000 polypropylene Griffin low-form beaker, 4000 mL WW-06000-80 Mfr # 1201-4000
COMPARE
4000 250 Polypropylene(PP)
₹15,900.00INR / Each
Showing 1- 10 of 10