Thermo Scientific™ Nalgene™ Polymethylpentene Griffin Low-Form Beakers
Don’t sacrifice performance
  • Get the transparency of glass and great chemical resistance
  • Autoclavable
Polymethylpentene (PMP) beakers offer the transparency of glass along with excellent chemical resistance. These beakers can be used continuously to 302°F (150°C) and intermittently to 347°F (175°C). Graduated with silk-screened markings, the graduations are easy-to-read.
12,000.00 - ₹23,700.00INR / Pkg of 12
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Thermo Scientific Nalgene 1203-0050 polymethylpentene Griffin low-form beaker, 50 mL WW-06001-87 Mfr # 1203-0050
COMPARE
50 10 Polymethylpentene
₹14,900.00INR / Pkg of 12
Thermo Scientific Nalgene 1203-0100 polymethylpentene Griffin low-form beaker, 100 mL WW-06001-88 Mfr # 1203-0100
COMPARE
100 10 Polymethylpentene
₹17,700.00INR / Pkg of 12
Thermo Scientific Nalgene 1203-0150 polymethylpentene Griffin low-form beaker, 150 mL WW-06001-89 Mfr # 1203-0150
COMPARE
150 10 Polymethylpentene
₹23,700.00INR / Pkg of 12
Thermo Scientific Nalgene 1203-0250 polymethylpentene Griffin low-form beaker, 250 mL WW-06001-90 Mfr # 1203-0250
COMPARE
250 25 Polymethylpentene
₹15,200.00INR / Pkg of 6
Thermo Scientific Nalgene 1203-0400 polymethylpentene Griffin low-form beaker, 400 mL WW-06001-91 Mfr # 1203-0400
COMPARE
400 25 Polymethylpentene
₹23,200.00INR / Pkg of 6
Thermo Scientific Nalgene 1203-0600 polymethylpentene Griffin low-form beaker, 600 mL WW-06001-92 Mfr # 1203-0600
COMPARE
600 25 Polymethylpentene
₹18,400.00INR / Pkg of 4
Thermo Scientific Nalgene 1203-1000 polymethylpentene Griffin low-form beaker, 1000 mL WW-06001-93 Mfr # 1203-1000
COMPARE
1000 50 Polymethylpentene
₹16,700.00INR / Pkg of 3
Thermo Scientific Nalgene 1203-2000 polymethylpentene Griffin low-form beaker, 2000 mL WW-06001-94 Mfr # 1203-2000
COMPARE
2000 100 Polymethylpentene
₹12,000.00INR / Each
Thermo Scientific Nalgene 1203-4000 polymethylpentene Griffin low-form beaker, 4000 mL WW-06001-95 Mfr # 1203-4000
COMPARE
4000 250 Polymethylpentene
₹18,900.00INR / Each
Showing 1- 9 of 9