Thermo Scientific Nalgene Durable Burettes (Burets) with Acrylic Body
Individually calibrated to meet ASTM E287 accuracy requirements
  • Won't break — durable burettes (burets) constructed of acrylic-body
Use these clear, acrylic-body burettes (Burets) with polymethylpentene tips and PTFE stopcocks over the entire pH range. Burettes are unaffected by dilute mineral acids and bases (except HF and ammonium hydroxide). The pH range of 1 to 14 is ideal and up to 5% wt/wt of acids or 30% bases can be used. Not designed for use with alcohols or organic solvents.
NOTE: Not designed for use with alcohols or organic solids
31,500.00 - ₹35,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Tolerance
Availability Pricing
Thermo Scientific Nalgene 3650-0025 Durable Acylic Body Burette, 25 mL; 1/Pk WW-06198-20 Mfr # 3650-0025
COMPARE
25 0.1 0.06
₹31,500.00INR / Each
Thermo Scientific Nalgene 3650-0050 Durable Acylic Body Burette, 50 mL; 1/Pk WW-06198-30 Mfr # 3650-0050
COMPARE
50 0.1 0.1
₹35,600.00INR / Each