Thermo Scientific Nalgene Analytical Funnels, PP
No air lock with outer ribs
  • Get faster filtration with 58° angled internal ribs
  • Accepts standard filter papers
  • Autoclavable
These funnels can use standard filter paper. Long stems and external ribbing eliminates air lock. The funnels are shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. The body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration.
9,600.00 - ₹14,200.00INR / Pkg of 4
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Filter Paper Diameter (mm) Top OD (mm)
Availability Pricing
Thermo Scientific Nalgene 4250-0035 polypropylene analytical funnel, 10 mL WW-06120-10 Mfr # 4250-0035
COMPARE
10 55 41
₹11,100.00INR / Pkg of 12
Thermo Scientific Nalgene 4250-0055 polypropylene analytical funnel, 41 mL WW-06120-20 Mfr # 4250-0055
COMPARE
41 90 61
₹12,000.00INR / Pkg of 12
Thermo Scientific Nalgene 4250-0065 polypropylene analytical funnel, 65 mL WW-06120-30 Mfr # 4250-0065
COMPARE
65 110 63
₹14,200.00INR / Pkg of 12
Thermo Scientific Nalgene 4250-0075 polypropylene analytical funnel, 114 mL WW-06120-40 Mfr # 4250-0075
COMPARE
114 125 84
₹10,100.00INR / Pkg of 6
Thermo Scientific Nalgene 4250-0100 polypropylene analytical funnel, 254 mL WW-06120-60 Mfr # 4250-0100
COMPARE
254 185 108
₹9,600.00INR / Pkg of 4
Thermo Scientific Nalgene 4250-0160 polypropylene analytical funnel, 976 mL WW-06120-70 Mfr # 4250-0160
COMPARE
976 240 166
₹11,600.00INR / Pkg of 2