This product is now discontinued, View Related Products
Thermo Scientific Mini Digital Orbital Shaker
Easy-to-use digitally-controlled shaker provides quiet horizontal mixing for a variety of vessels
  • PID control for a consistent smooth orbital motion
  • Compact design easily fits in incubators (non-CO2), chambers, and refrigerators (up to 60°C)
  • Digital speed control easily adjusts from 50 to 300 rpm
  • Use in continuous shaking or timer mode up to 99 hrs 59 min
  • Variety of accessories customize unit for exact laboratory needs
  • Must be ordered with platform (sold separately)
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Load (lbs) Max Speed (rpm) Power (VAC)
Availability Pricing
Thermo Scientific 88880025 Mini Digital Orbital Shaker, 100-240 VAC; US Plug WW-04725-14 Mfr # 88880025
COMPARE
8.8 300 100 to 240
Call For Price
Thermo Scientific 88880026 Mini Digital Orbital Shaker, 100-240 VAC; Int’nl Plug WW-04725-15 Mfr # 88880026
COMPARE
8.8 300 100 to 240