Thermo Scientific™ Merlin™ Recirculating Chillers
Precise temperature control in a compact design
  • Translucent reservoir lets you check liquid level and fluid condition at a glance
  • Digital display provides simple operation and easy-to-read measurements
  • Removable grill allows for quick and easy preventive maintenance
  • Global compliance ensures user safety
Compact, portable recirculating chillers are ideal for applications requiring heat removal across a wide temperature range. Offering two cooling capacities with a temperature range from –15°C to 35°C, the chiller platform provides powerful cooling in a reliable and compact design. The PWM valve regulates the amount of cooling required in relation to the amount of heat being removed, eliminating on/off cycling and premature wear of the compressor. Highly functional, configurable controller features a user-adjustable low- and high-temperature safety with audible alarm.
A variety of additional options—including low-level or low-flow indicators, remote start, and RS-232—let you configure these chillers for a wide range of application requirements.
1,562,000.00 - ₹2,805,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity At 20° C (watts) Power (Hz) Power (VAC)
Availability Pricing
Thermo Scientific Merlin 264216040002 Merlin 75  Low-Temperature Refrigerated Chillers, PD2 Pump, HFE Compatible; 230 VAC, 60 Hz WW-13401-51 Mfr # 264216040002
COMPARE
2156 60 230
₹2,805,000.00INR / Each
Thermo Scientific Merlin 264216044000 Merlin 75 Low-Temperature Refrigerated Chillers PD2 pump, with Low Level/Low Flow Switches, Remote Start and RS-232; 230 VAC, 60 Hz WW-13401-53 Mfr # 264216044000
COMPARE
2156 60 230
₹1,562,000.00INR / Each
Thermo Scientific Merlin 266216022000 Merlin 150 Low-Temperature Refrigerated Chillers, CP55 Pump, with Remote Start and RS-232; 230 VAC, 60 Hz WW-13401-65 Mfr # 266216022000
COMPARE
4832 60 230
₹2,367,000.00INR / Each
Thermo Scientific Merlin 266216042000 Merlin 150 Low-Temperature Refrigerated Chillers, PD2 Pump, with Remote Start and RS-232; 230 VAC, 60 Hz WW-13401-69 Mfr # 266216042000
COMPARE
4832 60 230
₹2,075,000.00INR / Each
Thermo Scientific Merlin 266216044000 Merlin 150 Low-Temperature Refrigerated Chillers, PD2 Pump, with Low Level/Low Flow Switches, Remote Start and RS-232; 230 VAC, 60 Hz WW-13401-71 Mfr # 266216044000
COMPARE
4832 60 230
₹2,251,000.00INR / Each