This product is now discontinued, View Related Products
Thermo Scientific MaxQ 4450 Benchtop Incubating Shakers
Small footprint requires minimal bench space
  • Simultaneously view speed, time, and temperature on digital model
Analog Shaker features a rotary dial with variable 40 to 400 rpm speed control for gentle stirring to vigorous agitation. Tachometer monitors and displays speed in rpm to guarantee accurate settings. Unit can be set for timed studies from 1 to 60 minutes or operate continuously.
Digital Shaker features variable 15 to 500 rpm speed control, using a membrane keypad with push-button controls. Individual displays for temperature, speed, and operating time minimize errors in establishing running conditions. Visual/audible signals alert operator to temperature deviations of ±1°C from set point. Unit will shut down and alert operator if rotation varies ±10% of set speed preventing shaker from walking. Soft start features elimina
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Thermo Scientific Analog 4450 Benchtop Incubating Shaker, , 120 VAC WW-51960-00 Mfr # SHKA4450
COMPARE
120
Call For Price
Thermo Scientific Analog 4450 Benchtop Incubating Shaker, , 240 VAC WW-51960-02 Mfr # SHKA4450-1CE
COMPARE
240
Call For Price
Thermo Scientific Digital 4450 Benchtop Incubating Shaker, , 120 VAC WW-51960-04 Mfr # SHKE4450
COMPARE
120
Call For Price
Thermo Scientific Digital 4450 Benchtop Incubating Shaker, , 240 VAC WW-51960-06 Mfr # SHKE4450-1CE
COMPARE
240