Thermo Scientific™ HyPURITY™ C18 Drop-In Guard Cartridges
Protect and prolong the life of your column
  • Easily trap contaminants such as solvent and sample debris
  • Convenient and economical
Drop-In Guard Cartridges will extend the life and performance of your column. These cartridges provide an economical and efficient way of trapping contaminants such as solvent and sample debris. Drop-in design makes for easy guard and column replacement—simply position between the injector and analytical column to remove damaging components before they reach the column. The 10-mm length offers maximum protection with minimal retention.
61,100.00INR / Pkg of 4
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Description
Availability Pricing
Thermo Scientific HyPurity Drop-In Guard Cartridges, C18, 3 µm, 10 mm L x 1.0mm ID; 4/Pk WW-01875-43 Mfr # 22103-011001
COMPARE
1 10 Drop-In Guard Cartridges, C18, 3 µm, 10 mm L x 1.0mm ID; 4/Pk
₹61,100.00INR / Pkg of 4