Thermo Scientific™ Hypersil™ SAS C1 Drop-In Guard Cartridges
Protect and prolong the life of your column
  • Easily trap contaminants such as solvent and sample debris
  • Convenient and economical
Drop-In Guard Cartridges will extend the life and performance of your column. These cartridges provide an economical and efficient way of trapping contaminants such as solvent and sample debris. Drop-in design makes for easy guard and column replacement — simply position between the injector and analytical column to remove damaging components before they reach the column. The 10-mm length offers maximum protection with minimal retention.
74,800.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil SAS Drop-In Guard Cartridges, C1, 5 µm, 10 mm L x 4.0 mm ID; 4/Pk WW-01888-59 Mfr # 30505-014001
COMPARE
4 5
₹74,800.00INR / Each