This product is now discontinued, View Related Products
Thermo Scientific Hypersil GOLD aQ Columns
Thermo Scientific Hypersil GOLD aQ Columns offer a polar endcapped C18 phase that provides the same optimized peak shape and resolution with the added selectivity and retention of polar compounds. Ideal for the quantitative analysis of polar analytes present in trace amounts and for reversed phase applications using 100% aqueous mobile phase. Stable in a pH range of 2 to 9.
Call For Price
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 25302-021030 GOLD aQ Column - Length 20mm, 1.0mm ID, 1.9µm particle size WW-01880-09 Mfr # 25302-021030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-022130 GOLD aQ Column - Length 20mm, 2.1mm ID, 1.9µm particle size WW-01880-10 Mfr # 25302-022130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-023030 GOLD aQ Column - Length 20mm, 3.0mm ID, 1.9µm particle size WW-01880-11 Mfr # 25302-023030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-031030 GOLD aQ Column - Length 30mm, 1.0mm ID, 1.9µm particle size WW-01880-12 Mfr # 25302-031030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-032130 GOLD aQ Column - Length 30mm, 2.1mm ID, 1.9µm particle size WW-01880-13 Mfr # 25302-032130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-033030 GOLD aQ Column - Length 30mm, 3.0mm ID, 1.9µm particle size WW-01880-14 Mfr # 25302-033030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-051030 GOLD aQ Column - Length 50mm, 1.0mm ID, 1.9µm particle size WW-01880-16 Mfr # 25302-051030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-052130 GOLD aQ Column - Length 50mm, 2.1mm ID, 1.9µm particle size WW-01880-19 Mfr # 25302-052130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-053030 GOLD aQ Column - Length 50mm, 3.0mm ID, 1.9µm particle size WW-01880-20 Mfr # 25302-053030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-101030 GOLD aQ Column - Length 100mm, 1.0mm ID, 1.9µm particle size WW-01880-26 Mfr # 25302-101030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-102130 GOLD aQ Column - Length 100mm, 2.1mm ID, 1.9µm particle size WW-01880-27 Mfr # 25302-102130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-103030 GOLD aQ Column - Length 100mm, 3.0mm ID, 1.9µm particle size WW-01880-28 Mfr # 25302-103030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-152130 GOLD aQ Column - Length 150mm, 2.1mm ID, 1.9µm particle size WW-01880-29 Mfr # 25302-152130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25302-202130 GOLD aQ Column - Length 200mm, 2.1mm ID, 1.9µm particle size WW-01880-30 Mfr # 25302-202130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25303-031030 GOLD aQ Column - Length 30mm, 1.0mm ID, 3µm particle size WW-01880-38 Mfr # 25303-031030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25303-032130 GOLD aQ Column - Length 30mm, 2.1mm ID, 3µm particle size WW-01880-39 Mfr # 25303-032130
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25303-033030 GOLD aQ Column - Length 30mm, 3.0mm ID, 3µm particle size WW-01880-40 Mfr # 25303-033030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25303-034030 GOLD aQ Column - Length 30mm, 4.0mm ID, 3µm particle size WW-01880-41 Mfr # 25303-034030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25303-034630 GOLD aQ Column - Length 30mm, 4.6mm ID, 3µm particle size WW-01880-42 Mfr # 25303-034630
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25303-051030 GOLD aQ Column - Length 50mm, 1.0mm ID, 3µm particle size WW-01880-46 Mfr # 25303-051030
COMPARE
3
Call For Price
Showing 1 - 20 of 50