This product is now discontinued, View Related Products
Thermo Scientific Cimarec External Remote Controllers
Compatible with Cimarec stirrers
7-Watt controller (04651-01, -03) is a compact controller ideal for small to medium volumes of low-viscosity liquids. It features continuous speed selection and automatic start-up for safe stirrer acceleration.
20-Watt controller (04651-04) is a menu-driven microprocessor controlled unit. The knob is easily controlled with one hand by pushing down and rotating. Recent settings are stored in the memory. These controllers feature three progam keys for individually programming rapid start up and a shake mode for vertical mixing.
40-Watt Controller (04651-05) offers all the same functions as the 20-Watt controller with a higher power output of 40 Watts for more demanding applications. This model has ten power settings and includes RS-232 interface for data transfer and PC control. Additionally, it has a TC-IP network integration option.
124,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Thermo Scientific Cimarec 50119115 i Telemodul controller 230V EU WW-04651-01 Mfr # 50119115
COMPARE
230
Call For Price
Thermo Scientific Cimarec 50119119 i Telemodul controller 115 US WW-04651-03 Mfr # 50119119
COMPARE
115
₹124,000.00INR / Each
Thermo Scientific Cimarec 50090773 Telemodul 20 C, 20 Watt Control Unit 100-240V WW-04651-04 Mfr # 50090773
COMPARE
100 to 240
Call For Price
Thermo Scientific Cimarec 50090774 Telemodul 40C, 40 Watt Control Unit 100-240V WW-04651-05 Mfr # 50090774
COMPARE
100 to 240