Thermo Scientific Bottle/Tube Roller
Compact, low-profile design and optional remote control make this ideal for use in incubators
  • Digitally controlled, reproducible rolling from 1 to 80 rpm
  • Remote control option lets you operate the roller unit from up to 9 ft (2.7 meters) away
  • Adjustable roller bars accommodate a variety of sizes of culture tubes and bottles
  • Save valuable bench space by stacking units two high with optional kit
Sleek, low-profile roller unit is versatile enough for multiple sizes of culture tubes and bottles. O-rings ensure smooth rolling of 15- and 50-mL tubes. Use the optional stacking kit to add on another roller unit and double your capacity, maximizing valuable bench or chamber space.
Robust, corrosion-resistant construction is safe enough to use in CO2 and humid environments. Order the optional remote controller to conveniently control the rolling speed and timer function without disturbing the incubating samples.
Order additional accessories such as stacking bars, additional O-rings, and remote control separately.
381,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
Thermo Scientific 88881003 Bottle/Tube Roller, 120V/US Plug WW-01836-03 Mfr # 88881003
COMPARE
120 Bottle/Tube Roller, 120V/US Plug
₹381,000.00INR / Each