This product is now discontinued, View Related Products

Thermo Scientific BioBasic SCX (Strong Cation Exchange) Columns

Stable polymeric coating suitable for separation and retention of basic and other cationic species
Thermo Scientific BioBasic HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic SCX Columns utilize a polymeric sulfonic acid ion exchange ligand that is bound to a highly pure, base-deactivated silica that creates a stable polymeric coating, even in demanding pH conditions (pH range of 2 to 8). Proven in the separation of cationic proteins and peptides, as well as neurotransmitters, vitamins, and quaternary amine herbicides. These columns are extremely reliable, providing reproducible results run after run.
Call For Price

20 variations of this product are available.

See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific BioBasic 73205-051030 SCX Column - Length 50mm, 1.0mm ID, 5µm particle size WW-01901-65 Mfr # 73205-051030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-052130 SCX Column - Length 50mm, 2.1mm ID, 5µm particle size WW-01901-68 Mfr # 73205-052130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-053030 SCX Column - Length 50mm, 3.0mm ID, 5µm particle size WW-01901-69 Mfr # 73205-053030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-054030 SCX Column - Length 50mm, 4.0mm ID, 5µm particle size WW-01901-70 Mfr # 73205-054030
COMPARE
4 50 5
Call For Price
Thermo Scientific BioBasic 73205-054630 SCX Column - Length 50mm, 4.6mm ID, 5µm particle size WW-01901-71 Mfr # 73205-054630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-101030 SCX Column - Length 100mm, 1.0mm ID, 5µm particle size WW-01901-81 Mfr # 73205-101030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-102130 SCX Column - Length 100mm, 2.1mm ID, 5µm particle size WW-01901-84 Mfr # 73205-102130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-103030 SCX Column - Length 100mm, 3.0mm ID, 5µm particle size WW-01901-85 Mfr # 73205-103030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-104030 SCX Column - Length 100mm, 4.0mm ID, 5µm particle size WW-01901-86 Mfr # 73205-104030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-104630 SCX Column - Length 100mm, 4.6mm ID, 5µm particle size WW-01901-87 Mfr # 73205-104630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-151030 SCX Column - Length 150mm, 1.0mm ID, 5µm particle size WW-01901-97 Mfr # 73205-151030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-152130 SCX Column - Length 150mm, 2.1mm ID, 5µm particle size WW-01901-98 Mfr # 73205-152130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-153030 SCX Column - Length 150mm, 3.0mm ID, 5µm particle size WW-01901-99 Mfr # 73205-153030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-154030 SCX Column - Length 150mm, 4.0mm ID, 5µm particle size WW-01902-00 Mfr # 73205-154030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-154630 SCX Column - Length 150mm, 4.6mm ID, 5µm particle size WW-01902-01 Mfr # 73205-154630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-251030 SCX Column - Length 250mm, 1.0mm ID, 5µm particle size WW-01902-02 Mfr # 73205-251030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-252130 SCX Column - Length 250mm, 2.1mm ID, 5µm particle size WW-01902-03 Mfr # 73205-252130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-253030 SCX Column - Length 250mm, 3.0mm ID, 5µm particle size WW-01902-04 Mfr # 73205-253030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-254030 SCX Column - Length 250mm, 4.0mm ID, 5µm particle size WW-01902-05 Mfr # 73205-254030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 73205-254630 SCX Column - Length 250mm, 4.6mm ID, 5µm particle size WW-01902-06 Mfr # 73205-254630
COMPARE
4.6 250 5
Call For Price
Showing 1- 20 of 20

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat