This product is now discontinued, View Related Products

Thermo Scientific BioBasic Phenyl Columns

Provide alternate selectivity, especially for peptides with aromatic side chains
  • Lower carbon loading for retention of larger proteins and peptides
Thermo Scientific BioBasic HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic Phenyl Columns contain 300Å pore size silica for maximum performance when used with biomolecules. These columns are ideally suited for the separation of high-hydrophobicity peptides nominally greater than 5,000 Da, and those with aromatic side chains. BioBasic Phenyl columns are also useful where alkyl chain chemistries do not provide adequate enough selectivity. The highly pure, deactivated silica in these columns contain extra dense bonding to provid
Call For Price

20 variations of this product are available.

See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific BioBasic 72405-051030 Phenyl Column - Length 50mm, 1.0mm ID, 5µm particle size WW-01899-03 Mfr # 72405-051030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-052130 Phenyl Column - Length 50mm, 2.1mm ID, 5µm particle size WW-01899-04 Mfr # 72405-052130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-053030 Phenyl Column - Length 50mm, 3.0mm ID, 5µm particle size WW-01899-05 Mfr # 72405-053030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-054030 Phenyl Column - Length 50mm, 4.0mm ID, 5µm particle size WW-01899-06 Mfr # 72405-054030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-054630 Phenyl Column - Length 50mm, 4.6mm ID, 5µm particle size WW-01899-07 Mfr # 72405-054630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-101030 Phenyl Column - Length 100mm, 1.0mm ID, 5µm particle size WW-01899-13 Mfr # 72405-101030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-102130 Phenyl Column - Length 100mm, 2.1mm ID, 5µm particle size WW-01899-14 Mfr # 72405-102130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-103030 Phenyl Column - Length 100mm, 3.0mm ID, 5µm particle size WW-01899-15 Mfr # 72405-103030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-104030 Phenyl Column - Length 100mm, 4.0mm ID, 5µm particle size WW-01899-16 Mfr # 72405-104030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-104630 Phenyl Column - Length 100mm, 4.6mm ID, 5µm particle size WW-01899-17 Mfr # 72405-104630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-151030 Phenyl Column - Length 150mm, 1.0mm ID, 5µm particle size WW-01899-18 Mfr # 72405-151030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-152130 Phenyl Column - Length 150mm, 2.1mm ID, 5µm particle size WW-01899-19 Mfr # 72405-152130
COMPARE
2.1 150 5
Call For Price
Thermo Scientific BioBasic 72405-153030 Phenyl Column - Length 150mm, 3.0mm ID, 5µm particle size WW-01899-20 Mfr # 72405-153030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-154030 Phenyl Column - Length 150mm, 4.0mm ID, 5µm particle size WW-01899-21 Mfr # 72405-154030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-154630 Phenyl Column - Length 150mm, 4.6mm ID, 5µm particle size WW-01899-22 Mfr # 72405-154630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-251030 Phenyl Column - Length 250mm, 1.0mm ID, 5µm particle size WW-01899-23 Mfr # 72405-251030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-252130 Phenyl Column - Length 250mm, 2.1mm ID, 5µm particle size WW-01899-24 Mfr # 72405-252130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-253030 Phenyl Column - Length 250mm, 3.0mm ID, 5µm particle size WW-01899-25 Mfr # 72405-253030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-254030 Phenyl Column - Length 250mm, 4.0mm ID, 5µm particle size WW-01899-26 Mfr # 72405-254030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72405-254630 Phenyl Column - Length 250mm, 4.6mm ID, 5µm particle size WW-01899-27 Mfr # 72405-254630
COMPARE
4.6 250 5
Call For Price
Showing 1- 20 of 20

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat