This product is now discontinued, View Related Products

Thermo Scientific BioBasic CN (Cyano) Columns

Provide outstanding reproducibility and efficiency
  • Lower carbon loading for retention of larger proteins and peptides
Thermo Scientific BioBasic HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic CN Columns contain 300Å pore size silica for maximum performance when used with biomolecules. These columns are ideally suited for the separation of high-hydrophobicity peptides nominally greater than 5,000 Da. BioBasic CN columns provide alternate selectivity and changes in elution order due to interactions through hydrogen bonding, dipole-dipole interactions, and dispersive interactions due to alkyl chain chemistries. The highly pure, deactivated sil
Call For Price

20 variations of this product are available.

See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific BioBasic 72905-051030 CN Column - Length 50mm, 1.0mm ID, 5µm particle size WW-01899-56 Mfr # 72905-051030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-052130 CN Column - Length 50mm, 2.1mm ID, 5µm particle size WW-01899-57 Mfr # 72905-052130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-053030 CN Column - Length 50mm, 3.0mm ID, 5µm particle size WW-01899-58 Mfr # 72905-053030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-054030 CN Column - Length 50mm, 4.0mm ID, 5µm particle size WW-01899-59 Mfr # 72905-054030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-054630 CN Column - Length 50mm, 4.6mm ID, 5µm particle size WW-01899-60 Mfr # 72905-054630
COMPARE
5 5
Call For Price
Thermo Scientific BioBasic 72905-101030 CN Column - Length 100mm, 1.0mm ID, 5µm particle size WW-01899-66 Mfr # 72905-101030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-102130 CN Column - Length 100mm, 2.1mm ID, 5µm particle size WW-01899-67 Mfr # 72905-102130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-103030 CN Column - Length 100mm, 3.0mm ID, 5µm particle size WW-01899-68 Mfr # 72905-103030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-104030 CN Column - Length 100mm, 4.0mm ID, 5µm particle size WW-01899-69 Mfr # 72905-104030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-104630 CN Column - Length 100mm, 4.6mm ID, 5µm particle size WW-01899-70 Mfr # 72905-104630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-151030 CN Column - Length 150mm, 1.0mm ID, 5µm particle size WW-01899-71 Mfr # 72905-151030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-152130 CN Column - Length 150mm, 2.1mm ID, 5µm particle size WW-01899-72 Mfr # 72905-152130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-153030 CN Column - Length 150mm, 3.0mm ID, 5µm particle size WW-01899-73 Mfr # 72905-153030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-154030 CN Column - Length 150mm, 4.0mm ID, 5µm particle size WW-01899-74 Mfr # 72905-154030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-154630 CN Column - Length 150mm, 4.6mm ID, 5µm particle size WW-01899-75 Mfr # 72905-154630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-251030 CN Column - Length 250mm, 1.0mm ID, 5µm particle size WW-01899-76 Mfr # 72905-251030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-252130 CN Column - Length 250mm, 2.1mm ID, 5µm particle size WW-01899-77 Mfr # 72905-252130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-253030 CN Column - Length 250mm, 3.0mm ID, 5µm particle size WW-01899-78 Mfr # 72905-253030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-254030 CN Column - Length 250mm, 4.0mm ID, 5µm particle size WW-01899-79 Mfr # 72905-254030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72905-254630 CN Column - Length 250mm, 4.6mm ID, 5µm particle size WW-01899-80 Mfr # 72905-254630
COMPARE
5
Call For Price
Showing 1- 20 of 20

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat