This product is now discontinued, View Related Products

Thermo Scientific™ BioBasic 8 Columns

Provide excellent separation of a wide range of peptides
  • Excellent starting column for peptide and protein separations
Thermo Scientific BioBasic HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.

BioBasic 8 Columns contain 300Å pore size silica, which is ideally suited for medium-hydrophobicity peptides nominally less than 20,000 Da. Providing less hydrophobic retention than BioBasic 18 columns, BioBasic 8 columns are great starting columns for protein separations and may provide changes in elution patterns. Columns provide a highly stable, reproducible surface with sharp peak shap
Call For Price

20 variations of this product are available.

See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific BioBasic 72205-051030 8 Column - Length 50mm, 1.0mm ID, 5µm particle size WW-01897-89 Mfr # 72205-051030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-052130 8 Column - Length 50mm, 2.1mm ID, 5µm particle size WW-01897-92 Mfr # 72205-052130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-053030 8 Column - Length 50mm, 3.0mm ID, 5µm particle size WW-01897-93 Mfr # 72205-053030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-054030 8 Column - Length 50mm, 4.0mm ID, 5µm particle size WW-01897-94 Mfr # 72205-054030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-054630 8 Column - Length 50mm, 4.6mm ID, 5µm particle size WW-01897-95 Mfr # 72205-054630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-101030 8 Column - Length 100mm, 1.0mm ID, 5µm particle size WW-01898-05 Mfr # 72205-101030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 8 Column - Length 100mm, 2.1mm ID, 5µm particle size WW-01898-08 Mfr # 72205-102130
COMPARE
2.1 100 5
Call For Price
Thermo Scientific BioBasic 72205-103030 8 Column - Length 100mm, 3.0mm ID, 5µm particle size WW-01898-10 Mfr # 72205-103030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-104030 8 Column - Length 100mm, 4.0mm ID, 5µm particle size WW-01898-11 Mfr # 72205-104030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-104630 8 Column - Length 100mm, 4.6mm ID, 5µm particle size WW-01898-12 Mfr # 72205-104630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-151030 8 Column - Length 150mm, 1.0mm ID, 5µm particle size WW-01898-23 Mfr # 72205-151030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-152130 8 Column - Length 150mm, 2.1mm ID, 5µm particle size WW-01898-24 Mfr # 72205-152130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-153030 8 Column - Length 150mm, 3.0mm ID, 5µm particle size WW-01898-26 Mfr # 72205-153030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-154030 8 Column - Length 150mm, 4.0mm ID, 5µm particle size WW-01898-27 Mfr # 72205-154030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-154630 8 Column - Length 150mm, 4.6mm ID, 5µm particle size WW-01898-28 Mfr # 72205-154630
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-251030 8 Column - Length 250mm, 1.0mm ID, 5µm particle size WW-01898-30 Mfr # 72205-251030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-252130 8 Column - Length 250mm, 2.1mm ID, 5µm particle size WW-01898-31 Mfr # 72205-252130
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-253030 8 Column - Length 250mm, 3.0mm ID, 5µm particle size WW-01898-33 Mfr # 72205-253030
COMPARE
5
Call For Price
Thermo Scientific BioBasic 72205-254030 8 Column - Length 250mm, 4.0mm ID, 5µm particle size WW-01898-34 Mfr # 72205-254030
COMPARE
4 250 5
Call For Price
Thermo Scientific BioBasic 72205-254630 8 Column - Length 250mm, 4.6mm ID, 5µm particle size WW-01898-35 Mfr # 72205-254630
COMPARE
5
Call For Price
Showing 1- 20 of 20

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat