Thermo Scientific™ BioBasic™ 18 HPLC Columns
Provide excellent separation of small to medium peptides
  • High purity silica and stable bonding chemistry — ideal for Life Science applications
  • 300Å pore size for maximum performance with biomolecules
  • High peak capacity stationary phase
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Thermo Scientific™ BioBasic™ HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic 18 Columns provide reversed phase separation of small to medium peptides. Columns contain 300Å pore size silica, which is ideally suited for medium- to low-hydrophobicity peptides less than 5000 Da. BioBasic 18 HPLC columns provide a highly stable, reproducible surface with sharp peak shape due to the dense bonding chemistry and surface deactivation.
166,000.00 - ₹174,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific BioBasic 18 HPLC Column, C18, 5 µm, 150 mm L x 4.6 mm ID WW-01897-59 Mfr # 72105-154630
COMPARE
4.6 150 5
₹166,000.00INR / Each
Thermo Scientific BioBasic 18 HPLC Column, C18, 5 µm, 250 mm L x 4.6 mm ID WW-01897-69 Mfr # 72105-254630
COMPARE
4.6 250 5
₹174,000.00INR / Each