This product is now discontinued, View Related Products
Thermo Scientific AquaMate Spectrophotometers
Designed for water and wastewater analysis
  • Over 260 pre-programmed water quality methods plus user customizable methods
  • Methods stored on USB stick to be added or removed as needed
  • One-point calibration adjustment ensures the highest accuracy
  • Compatible with several leading brands of reagents
AquaMate spectrophotometers offer the accuracy, dependability and ease of use your lab requires. Compact design with intuitive operation targeted to water and wastewater analysis. The pre-programmed or user-customizable water quality methods offer absorbance, % transmittance and concentration testing. Ensure wavelength accuracy and quality measurements with performance verification testing in accordance to GLP and GMP.
Aquamate features three USB connectors, providing the option to store methods and data to a USB stick, transfer data to a computer, or print data to an external printer. A wide range of vial sizes can be used with the 3 position cell-holder (turret): 13 to 25 mm round vials, 20 to 50 mm rectangular vials, and a 10 mm square vial. System is compatible with Orion, Merck, Chemetrics, and Hach chemistries.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Bandwidth Max Wavelength (nm) Min Wavelength (nm)
Availability Pricing
Thermo Scientific 7000 Aquamate Vis Spectrometer WW-99564-10 Mfr # AQ7000
COMPARE
5.0 nm 1100 325
Call For Price
Thermo Scientific 8000 Aquamate Uv-Vis Spectrometer WW-99564-20 Mfr # AQ8000
COMPARE
1.8 nm 1100 190