Thermo Fisher Molecular BioProducts Sterile, DNase-, RNase-, Pyrogen-Free Pipette Tips
Pipette tips are certified sterile, DNase-, RNase-, and pyrogen-free to prevent sample contamination
  • Reference mark at 10 µL
  • Simplify loading and ejecting
Sterile, crystal clear tips let you see transparent samples. The graduated 200 µL tips have a reference mark at 10 µL so you can check pipettor calibration and visually estimate sample size. Soft-fit design of 1000 µL tips simplify loading and ejecting. Tips range in volume from 0.1 to 1000 µL and fit a variety of pipettors to ensure laboratory standardization.
21,400.00INR / Pkg of 10
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Package Of
Availability Pricing
Thermo Fisher Molecular BioProducts 110-96RNS MBP Pipette Tips, DNase/RNase/Pyrogen-Free, 200 uL; 960/Pk WW-25713-44 Mfr # 110-96RNS
COMPARE
200 Racks
₹21,400.00INR / Pkg of 10