Testo Thermocouple Thermometer with Wireless Capability
Professional and reliable, measure several temperatures simultaneously
  • Meter requires the probes to be ordered separately from an impressive range of options
  • On-site documentation is provided by the optional IR printer
  • Optional IP65-rated case keeps dirt away from the meter
  • Uses only Type K thermocouples
These robust, reliable meters work in tough environments. The sturdy construction protects against knocks and jars. The large illuminated display is slightly set back in the housing for better protection. The carrying strap is designed for safe transport and the magnets on the back of the unit provide a secure means of attachment to a metal object. The display of min or max values is possible for all temperature channels. Easy-to-use menu offer such functions as max/min value, Hold, audible alarm settings, and basic status such as battery life. These meters have an impressive range of optional probes (meter does not come with a probe) that can measure temperature within seconds. In addition to standard probes with a wire, this meter can be easily upgraded anytime to wireless probes for use up to a distance of 20 meters. To add this feature, order wireless upgrade kit 37486-02 (includes one wireless handle and module) along with the wireless thermocouple adapter 37486-03.
47,800.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° C) Number Of Probe Channels
Availability Pricing
Testo 0563 0922 Thermocouple Thermometer with Wireless Capability; 2 Channel, Type K WW-37486-16 Mfr # 0563 0922
COMPARE
1832 -50 2
₹47,800.00INR / Each