Testo Humidity Sticks
Ideal for quick monitoring of ambient conditions in hard-to-reach locations
  • Head swivels 180°— allows easy viewing at any angle
These humidity sticks are small enough to fit in your pocket. They feature a single button to switch from temperature, humidity and dew point or wet bulb. The water resistant sensor protects from condensation, and a sensor cap rotates to protect from damage. They also feature auto-off to preserve battery life.
Whats included: thermohygrometer stick with multi-purpose clip, duct holder, and battery.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Max RH Range (%) Min RH Range (%)
Availability Pricing
Testo 0560 6053 Humidity Stick with Dew Point WW-37461-00 Mfr # 0560 6053
COMPARE
3 AAA (included) 95 5