Chemware® PFA Test Tubes
View contents with ease
  • Low metal ion content test tubes
  • Constructed of chemically inert and noncontaminating PFA
  • Autoclavable
Chemware® PFA test tubes are ideal for trace metal/ICP analysis and applications requiring excellent chemical resistance. PFA has the lowest metal ion content and widest temperature range of any fluoropolymer resin. Test tubes are translucent for easy viewing of contents and withstand temperatures up to 260°C (500°F).
17,100.00 - ₹48,000.00INR / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Bottom Shape Tube Capacity (mL) Tube Material
Availability Pricing
Chemware D1069598 PFA Test Tubes with Caps, 5 mL, 10 x 120 mm; 1/Pk WW-63614-20 Mfr # D1069598
COMPARE
Round 5 PFA
₹17,100.00INR / Pkg of 1
Chemware D1069688 PFA Test Tubes with Caps, 10 mL, 12 x 140 mm; 1/Pk WW-63614-22 Mfr # D1069688
COMPARE
Round 10 PFA
₹18,700.00INR / Pkg of 1
Chemware D1069599 PFA Test Tubes with Caps, 15 mL, 16 x 110 mm; 1/Pk WW-63614-24 Mfr # D1069599
COMPARE
Round 15 PFA
₹21,900.00INR / Pkg of 1
Chemware D1069690 PFA Test Tubes with Caps, 25 mL, 19 x 150 mm; 1/Pk WW-63614-28 Mfr # D1069690
COMPARE
Round 25 PFA
₹29,700.00INR / Pkg of 1
Chemware D1069691 PFA Test Tube with Cap, 50 mL, 22 x 220 mm; 1/Pk WW-63614-30 Mfr # D1069691
COMPARE
Round 50 PFA
₹43,500.00INR / Pkg of 1
Chemware D1069692 PFA Test Tubes with Caps, 60 mL, 22 x 230 mm; 1/Pk WW-63614-32 Mfr # D1069692
COMPARE
Round 60 PFA
₹48,000.00INR / Pkg of 1