This product is now discontinued, View Related Products
Techne Systemic Viral Pathogen Detection Kits for qPCR
Quickly screen for viral pathogens to prevent the spreading of disease causing systemic viruses
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Prevent economic loss, especially in the dairy and agricultural industry, using these viral detection kits to accurately assess whether these disease-causing pathogens are present. Select a complete kit with mastermix or order a kit without mastermix and use your own.
Call For Price
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Mastermix Included Test Type Description
Availability Pricing
COMPARE
No RNA Avian Orthoreovirus, without mastermix
Call For Price
COMPARE
Yes RNA Avian Orthoreovirus, with mastermix
Call For Price
COMPARE
No RNA African Horse Sickness Virus, without mastermix
Call For Price
COMPARE
Yes RNA African Horse Sickness Virus, with mastermix
Call For Price
COMPARE
No DNA Aleutian Disease Virus, without mastermix
Call For Price
COMPARE
Yes DNA Aleutian Disease Virus, with mastermix
Call For Price
COMPARE
No DNA Budgerigar Fledgling Disease Virus (avian polyomavirus), without mastermix
Call For Price
COMPARE
Yes DNA Budgerigar Fledgling Disease Virus (avian polyomavirus), with mastermix
Call For Price
COMPARE
No RNA Bundibugyo ebolavirus, without mastermix
Call For Price
COMPARE
Yes RNA Bundibugyo ebolavirus, with mastermix
Call For Price
COMPARE
No DNA Beak and Feather Disease Virus, without mastermix
Call For Price
COMPARE
Yes DNA Beak and Feather Disease Virus, with mastermix
Call For Price
COMPARE
No DNA Bovine Herpesvirus 1, without mastermix
Call For Price
COMPARE
Yes DNA Bovine Herpesvirus 1, with mastermix
Call For Price
COMPARE
No RNA Bovine Leukemia Virus, without mastermix
Call For Price
COMPARE
Yes RNA Bovine Leukemia Virus, with mastermix
Call For Price
COMPARE
No RNA Bluetongue Virus, without mastermix
Call For Price
COMPARE
No RNA Bluetongue Virus 1, without mastermix
Call For Price
COMPARE
Yes RNA Bluetongue Virus 1, with mastermix
Call For Price
COMPARE
No RNA Bluetongue Virus 8, without mastermix
Call For Price
Showing 1 - 20 of 50