Cole-Parmer® Replacement Spinner Flasks for BS-200 Cell Culture Stirring Systems
Replacement cell culture glass spinner flask for BS-200 cell culture stirring systems
  • Unique protrusion in base allows for gentle mixing to help eliminate cell damage
  • Silicone-encapsulated to prevent cells from attaching and clumping
  • Constructed of borosilicate glass
These borosilicate glass replacement spinner flasks are for use with caps and stirring mechanisms from Cole-Parmer® spinner flask assemblies. They incorporate a unique base design, which in cooperation with the bulb-ended glass stirrer rod (not included), ensures that cultures are lifted into suspension at the lowest possible speeds. The flask is silicone-encapsulated to reduce the possibility of cells attaching to and growing on the surfaces. Culture vessels come calibrated, and have two side ports and a central filling port. They are suitable for repeated autoclaving at 121 °C (250 °F) and 15 psi for 30 minutes. These flasks do not include caps or stirring mechanisms and are intended as replacement flasks only.
29,700.00 - ₹59,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Height (cm) Description
Availability Pricing
COMPARE
500 20.5 Replacement Spinner Flask Only for BS-200 Cell Culture Stirring Systems, 500 mL (16.9 oz)
₹39,700.00INR / Each
COMPARE
1000 26.3 Replacement Spinner Flask Only for BS-200 Cell Culture Stirring Systems, 1000 mL (33.8 oz)
₹59,400.00INR / Each
COMPARE
250 17 Replacement Spinner Flask Only for BS-200 Cell Culture Stirring Systems, 250 mL (8.5 oz)
₹29,700.00INR / Each
COMPARE
125 14.5 Replacement Spinner Flask Only for BS-200 Cell Culture Stirring Systems, 125 mL (4.2 oz)
₹29,700.00INR / Each