This product is now discontinued, View Related Products
Techne­ Prime Large-Format Thermal Cyclers
Choose from a reliable and robust selection of thermal cycling options based on your needs
  • Colorful touch screen allows for quick set up and easy programming with intuitive software
  • Uses the Peltier effect—a chosen method for the most trusted thermal cyclers on the market
  • Heated lid prevents condensation in your samples
  • Easily upgrade your unit to gradient cycling, now or later
These high-performance thermal cyclers provide robust and reliable operation that is user friendly and easy on the pocketbook. Intuitive programming feature allows you to easily set up to 1000 programs (unlimited with USB) with password protection to minimize inadvertent changes in settings. Use the color touch screen to create even the most complex programs.
Heated lid has an adjustable temperature range of 100 to 115°C to prevent condensation in your samples. All units have the ability to transfer programs via the USB port between instruments or to your PC. An industry-leading 4-year warranty ensures years of reliable results.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Heating Rate Temperature Accuracy Description
Availability Pricing
COMPARE
up to 3.4°C/sec ±0.2°C at 50°C Thermal Cycler, 96 x 0.2 mL
Call For Price
COMPARE
up to 3.4°C/sec ±0.2°C at 50°C Thermal Cycler, 60 X 0.5 mL
Call For Price
COMPARE
up to 3.4°C/sec ±0.2°C at 50°C Thermal Cycler, 384 Well Plates
Call For Price
COMPARE
up to 3.4°C/sec ±0.2°C at 50°C Gradient Thermal Cycler, 96 x 0.2 mL
Call For Price
COMPARE
up to 3.4°C/sec ±0.2°C at 50°C Gradient Thermal Cycler, 60 x 0.5 mL
Call For Price
COMPARE
up to 3.4°C/sec ±0.2°C at 50°C Gradient Thermal Cycler, 384 Wells