Cole-Parmer® IC-300 Series High-Temperature , High-Stability Digital Immersion Circulator
Accurately control liquid bath temperature from -40 to 250°C
  • Precise PID temperature control with temperature stability of ±0.005°C
  • Bidirectional RS-232 interface
This versatile immersion circulator can be used in several ways. Use as a stand-alone water bath temperature controller or a pump for recirculating chilled liquid from an external chiller. Immersion circulator accurately heats up to 250°C and safely controls the temperature of liquid in the bath while maintaining excellent temperature stability. A bidirectional RS-232 interface lets you control bath remotely and download data to a computer for further analysis.
Precise PID temperature control, a bright digital LED display, overtemperature indicator light, pressure pump, and low-liquid level cutoff make the curculator easy and safe to use. Reach below ambient temperatures by using the cooling coil 12122-87 connected to an optional external chiller.
346,000.00 - ₹398,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
120 High-Temperature, High-Stability Digital Immersion Circulator; 120 VAC, 50/60 Hz
₹346,000.00INR / Each
COMPARE
230 High-Temperature, High-Stability Digital Immersion Circulator; 230 VAC, 50/60 Hz
₹398,000.00INR / Each