This product is now discontinued, View Related Products
Techne Human Viral Pathogen Detection Kits for qPCR
Crucial tool for fast and accurate detection of dangerous human viral pathogens
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Call For Price
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Mastermix Included Test Type Description
Availability Pricing
COMPARE
No DNA Adenovirus type B, without mastermix
Call For Price
COMPARE
Yes DNA Adenovirus type B, with mastermix
Call For Price
COMPARE
No DNA Adenovirus type C, without mastermix
Call For Price
COMPARE
Yes DNA Adenovirus type C, with mastermix
Call For Price
COMPARE
No DNA Adenovirus type F&G, without mastermix
Call For Price
COMPARE
Yes DNA Adenovirus type F&G, with mastermix
Call For Price
COMPARE
No DNA BK Virus, without mastermix
Call For Price
COMPARE
Yes DNA BK Virus, with mastermix
Call For Price
COMPARE
No RNA Chaoyang Virus, without mastermix
Call For Price
COMPARE
Yes RNA Chaoyang Virus, with mastermix
Call For Price
COMPARE
No RNA Chikungunya Virus, without mastermix
Call For Price
COMPARE
Yes RNA Chikungunya Virus, with mastermix
Call For Price
COMPARE
No DNA Cytomegalovirus (HHV5), without mastermix
Call For Price
COMPARE
Yes DNA Cytomegalovirus (HHV5), with mastermix
Call For Price
COMPARE
No RNA Dengue Virus, without mastermix
Call For Price
COMPARE
No RNA Dengue Virus type 3, without mastermix
Call For Price
COMPARE
Yes RNA Dengue Virus type 3, with mastermix
Call For Price
COMPARE
Yes RNA Dengue Virus, with mastermix
Call For Price
COMPARE
No DNA Epstein Barr Virus (HHV4), without mastermix
Call For Price
COMPARE
Yes DNA Epstein Barr Virus (HHV4), with mastermix
Call For Price
Showing 1 - 20 of 50