This product is now discontinued, View Related Products
Techne Human Microbiome Kits for qPCR
Designed for the in vitro quantification of human gastrointestinal microbiome
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is specifically designed to ensure the broadest possible detection profile and to detect all relevant strains and subtypes. All kits are available with onestep mastermix or without.
156,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Mastermix Included Test Type Description
Availability Pricing
COMPARE
No DNA Bifidobacterium bifidum, without mastermix
Call For Price
COMPARE
Yes DNA Bifidobacterium bifidum, with mastermix
Call For Price
COMPARE
No DNA Bifidobacterium longum, without mastermix
Call For Price
COMPARE
Yes DNA Bifidobacterium longum, with mastermix
Call For Price
COMPARE
No DNA Lactobacillus plantarum, without mastermix
Call For Price
COMPARE
Yes DNA Lactobacillus plantarum, with mastermix
₹156,000.00INR / Each