This product is now discontinued, View Related Products
Techne Genetically Modified Organism Detection Kits for qPCR
Screen for genetically modified organisms (GMOs) in food, feed, and seeds
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Kits with mastermix contain all the necessary components for fast and accurate results. To use your own mastermix, order kits without mastermix.
Call For Price
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Mastermix Included Test Type Description
Availability Pricing
COMPARE
No DNA Genesig FMV 35S promoter in GM crops, without mastermix
Call For Price
COMPARE
Yes DNA Genesig FMV 35S promoter in GM crops, with mastermix
Call For Price
COMPARE
No DNA Genesig GMO integration event Bt11 in Maize, without mastermix
Call For Price
COMPARE
Yes DNA Genesig GMO integration event Bt11 in Maize, with mastermix
Call For Price
COMPARE
No DNA Genesig GMO integration event Bt176 in Maize, without mastermix
Call For Price
COMPARE
Yes DNA Genesig GMO integration event Bt176 in Maize, with mastermix
Call For Price
COMPARE
No DNA Genesig GMO integration event MON810 in Maize, without mastermix
Call For Price
COMPARE
Yes DNA Genesig GMO integration event MON810 in Maize, with mastermix
Call For Price
COMPARE
No DNA Genesig GMO integration event NK603 in Maize, without mastermix
Call For Price
COMPARE
Yes DNA Genesig GMO integration event NK603 in Maize, with mastermix
Call For Price
COMPARE
No DNA Genesig CaMV 35S promoter and NOS terminator in GM Maize, without mastermix
Call For Price
COMPARE
Yes DNA Genesig CaMV 35S promoter and NOS terminator in GM Maize, with mastermix
Call For Price
COMPARE
No DNA Genesig CaMV 35S promoter in GM crops, without mastermix
Call For Price
COMPARE
Yes DNA Genesig CaMV 35S promoter in GM crops, with mastermix
Call For Price
COMPARE
No DNA Genesig CaMV 35S promoter and NOS terminator in GM Soya, without mastermix
Call For Price
COMPARE
Yes DNA Genesig CaMV 35S promoter and NOS terminator in GM Soya, with mastermix
Call For Price
COMPARE
No DNA Genesig GMO integration event GTS 40-30-2 (Round up Ready Soya), without mastermix
Call For Price
COMPARE
Yes DNA Genesig GMO integration event GTS 40-30-2 (Round up Ready Soya), with mastermix
Call For Price
COMPARE
No DNA Genesig NOS terminator in GM crops, without mastermix
Call For Price
COMPARE
Yes DNA Genesig NOS terminator in GM crops, with mastermix
Call For Price
Showing 1- 20 of 20