This product is now discontinued, View Related Products

Techne Flow Coolers

Designed for use when temperatures down to -35°C are required
  • Compact refrigeration units with a 200 mL internal capacity
  • Designed for use with Techne liquid baths
The Techne® Flow Coolers work in conjunction with a Techne or Cole-Parmer® IC-300 series immersion circulator. The bath liquid flows through the unit which continually extracts heat from the bath fluid by means of the heat exchanger which is built into the unit.
526,000.00 - ₹856,000.00INR / Each

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Compatible With Power (VAC) Description
Availability Pricing
COMPARE
Techne Calibration Baths 115 Flow Cooler, -35⁰C; 115VAC
₹856,000.00INR / Each
COMPARE
Techne Calibration Baths 230 Flow Cooler, -20⁰C; 230 VAC
₹526,000.00INR / Each
COMPARE
Techne Calibration Baths 120 Flow Cooler, -20⁰C; 120 VAC
₹574,000.00INR / Each
COMPARE
Techne Calibration Baths 230 Flow Cooler, -35⁰C; 230 VAC/UK
Call For Price
COMPARE
Techne Calibration Baths 230 Flow Cooler, -35⁰C; 230 VAC/EU
Call For Price
COMPARE
External Bypass for Circulators
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat