Cole-Parmer HBC-300 Series Dry Block Temperature Calibrators
Fast heat up and cool down
  • Built rugged for portable field use
  • Ten available insert formats
  • Temperature sensor burnout protection
  • Lightweight, only 9 lbs
These low cost, protable dry block calibrators feature a wide temperature range with an independent over temperature cutout, and offer a maximum temperature of 430°C (806°F).
379,000.00 - ₹382,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Power (VAC) Temperature Accuracy
Availability Pricing
COMPARE
806 120 ±0.3°C at 200°C, ±0.5°C at 400°C
₹382,000.00INR / Each
COMPARE
806 240 ±0.3°C at 200°C, ±0.5°C at 400°C
₹379,000.00INR / Each