Block Heat Inserts for Cole-Parmer® BH-200 and BH-250 Series Block Heaters and CB-200 Series Chiller/Heater
Interchangeable heating blocks offer excellent thermal conductivity
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Depending on the combination of blocks selected, plastic spacers (36620-67) can be used to fill a half-width recess, ensuring optimal thermal performance. Most blocks come with a separate hole to accept a thermometer.
Order a flexi-block to accommodate multiple size tubes, vials, or cuvettes at once. Unskirted PCR plates can be heated in either size flexi-block. Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version.
5,400.00 - ₹97,200.00INR / Each
40 variations of this product are available.
See All Product Options Below
40 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Block Type Number Of Blocks Description
Availability Pricing
COMPARE
Ceramic beads/Aluminum 2 Block Heater Insert, Double Wide, Flexi-Block, Aluminum
₹34,100.00INR / Each
COMPARE
Ceramic beads/Aluminum 3 Block Heater Insert, Triple Wide, Flexi-Block, Aluminum
₹60,500.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Solid Aluminum
₹17,400.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, 12 mm dia x 33 mm Deep
₹7,900.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, 12 mm dia Holes
₹22,700.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, 17 mm dia Holes
₹18,700.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, 19 mm dia Holes
₹8,100.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, 20 x 2 mL Tubes
₹8,400.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, Cuvette and Pipette Channels
₹19,200.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, 3 x 4 Array, 5 mL Tubes
₹12,200.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, 25 mm dia Holes
₹19,000.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, 28 mm dia Holes
₹19,000.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, 33 mm dia Holes
₹19,400.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, 48 x 0.2 mL Tubes
₹22,800.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Single Wide, Aluminum, 80 x 0.2 mL Tube Strips
₹28,200.00INR / Each
COMPARE
Aluminum 2 Block Heater Insert, Double Wide, Aluminum, 96 Well Plate
₹62,800.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Conical Bottom, Aluminum, 30 x 6 mm dia
₹19,600.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Conical Bottom, Aluminum, 20 x 10 mm dia
₹14,900.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Conical Bottom, Aluminum, 20 x 12 mm dia
₹14,900.00INR / Each
COMPARE
Aluminum 1 Block Heater Insert, Conical Bottom, Aluminum, 20 x 13 mm dia
₹6,300.00INR / Each
Showing 1 - 20 of 40