Cole-Parmer® BH-250 Series Dry-Block Heaters
Choose a heater, then blocks to suit your application needs!
  • Precision-engineered heaters with excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Take up less storage space—lightweight and flat-top surface allow units to be stacked when not in use
Use these heaters as a dry, constant temperature source for various lab and research applications. All models have independent indicator lights to show main power, heater action, and over-temperature cut out. The “number of blocks accepted” on the ordering table indicate the number of single-width blocks accepted by each block heater. A heater that accepts two single-width blocks will accept one double-width block. Use three- and four-block models for high-throughput applications such as chemical oxygen demand (COD).
137,000.00 - ₹240,000.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Control Type Number Of Blocks Power (VAC)
Availability Pricing
COMPARE
Digital; 100°C 2 230
₹138,000.00INR / Each
COMPARE
Digital; 100°C 2 115
₹137,000.00INR / Each
COMPARE
Digital, Twin control; 100°C 2 230
₹214,000.00INR / Each
COMPARE
Digital, Twin control; 100°C 2 115
₹214,000.00INR / Each
COMPARE
Digital; 100°C 3 230
₹240,000.00INR / Each
COMPARE
Digital; 100°C 3 115
₹149,000.00INR / Each
COMPARE
Digital; 100°C 4 230
₹179,000.00INR / Each
COMPARE
Digital; 100°C 4 115
₹179,000.00INR / Each
COMPARE
Digital; 200°C 2 230
₹143,000.00INR / Each
COMPARE
Digital; 200°C 2 115
₹142,000.00INR / Each
COMPARE
Digital; 200°C 3 230
₹151,000.00INR / Each
COMPARE
Digital; 200°C 3 115
₹150,000.00INR / Each
COMPARE
Analog; 100°C 2 230
₹146,000.00INR / Each
COMPARE
Analog; 100°C 2 115
₹147,000.00INR / Each
COMPARE
Analog; 200°C 3 230
₹190,000.00INR / Each
COMPARE
Analog; 200°C 3 115
₹189,000.00INR / Each
COMPARE
Digital, RS-232; 200°C 3 230
₹217,000.00INR / Each
COMPARE
Digital, RS-232; 200°C 2 230
₹213,000.00INR / Each
COMPARE
Analog; 100°C 3 115
₹153,000.00INR / Each
Showing 1- 19 of 19