Cole-Parmer® Spinner Flask Assemblies for BS-200 Cell Culture Stirring Systems
A complete cell culture spinner flask for use with the BS-200 series stirring systems
  • Features a gentle stirring action to promote a high plating efficiency and high cell yield
  • Completely sealed vessel is ideal for pathogenic materials
  • Unique protrusion in base allows for gentle mixing to help eliminate cell damage
  • Silicone-encapsulated culture vessel and stir rod prevents cells from attaching and clumping
  • Includes flask, stir rod, and caps
Cole-Parmer® cell culture spinner flasks incorporate a unique upwards protrusion in the base that, in cooperation with the bulb-ended glass stirrer rod, helps lift cells into suspension at the lowest possible speeds. This aids attachment and promotes circulation of the medium around the cells to encourage a high rate of growth while helping to eliminate cell damage.
Both the culture vessel and stir rod are silicone-encapsulated to reduce the possibility of cells attaching to and growing on the surfaces. These borosilicate glass culture flasks come calibrated, and have two side ports and a central filling port. All culture vessel parts are suitable for repeated autoclaving at 121 °C (250 °F) and 15 psi for 30 minutes. All internal components are biologically inert.
59,400.00 - ₹94,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Height (cm) Description
Availability Pricing
COMPARE
500 20.5 Spinner Flask Assembly for BS-200 Cell Culture Stirring Systems, 500 mL (16.9 oz)
₹66,100.00INR / Each
COMPARE
1000 26.3 Spinner Flask Assembly for BS-200 Cell Culture Stirring Systems, 1000 mL (33.8 oz)
₹94,900.00INR / Each
COMPARE
250 17 Spinner Flask Assembly for BS-200 Cell Culture Stirring Systems, 250 mL (8.5 oz)
₹59,400.00INR / Each
COMPARE
125 14.5 Spinner Flask Assembly for BS-200 Cell Culture Stirring Systems, 125 mL (4.2 oz)
₹59,400.00INR / Each