This product is now discontinued, View Related Products
Techne 2-Position Dri-Block Heater with Two FREE Blocks
Precision-engineered heaters with excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Take up less storage space — lightweight and flat-top surface allow units to be stacked when not in use
  • Two FREE insert blocks included
Use these heaters as a dry, constant temperature source for various lab and research applications. All models have independent indicator lights to show main power, heater action, and over-temperature cut out.
152,000.00 - ₹155,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
2-Position Heating Block with Two Free Blocks, 20 x 1.5 mL and 30 x 0.5 mL, 120 V
₹155,000.00INR / Each
COMPARE
2-Position Heating Block with Two Free Blocks, two 20 x 1.5 mL, 120 V
₹152,000.00INR / Each