Low-foaming, three-dimensional “wave” motion is ideal for a wide range of laboratory applications
- Easy-to-read, independent LED displays for speed/tilt angle and time allow you to view all settings at once
This shaker provides precise speed control and a smooth vertical and horizontal ''wave'' motion which is suitable for liquid samples, DNA extractions, blotting techniques, and general mixing of various-sized tubes. Cast-aluminum base offers durability and stability. Triple eccentric drive contains permanently lubricated ball bearings and a brushless DC motor for reliable service.
Digital LED display provides repeatable and accurate results every time. Timer displays elapsed time, user-defined limit, and will shut off unit when time reaches zero. Display shows last settings, even after power has been turned off. Audible and visual signals activate when the system detects an obstruction or overload of the tray for added safety.