This product is now discontinued, View Related Products
Foxx Life Sciences Syringe Filters, Sterile
Individually-wrapped, sterile sample filters
  • Reduce bursting with reinforced polypropylene housing
  • Take advantage of a large effective filtration area
  • Standard female luer-lock inlet and male luer-slip outlet
  • Made with USP Class VI components to ensure quality
  • Available in various membrane size, pore size, and material combinations
Use the single-use syringe filters for clarification and prefiltration, and even for removing fine particulate with the membrane’s large effective filtration area. Reinforced polypropylene housing has secure connections for male luer slip outlet and female luer lock inlet. Sterile syringe filters are individually packaged in an easy-open blister pack. The membrane type and pore size are visible on each filter housing as well.
39,400.00 - ₹39,900.00INR / Pkg of 100
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Membrane Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
COMPARE
13 PVDF 0.22
₹39,900.00INR / Pkg of 100
Foxx Life Sciences Syringe Filter, 25 mm, 0.22 um PVDF, Sterile; 100/Pk WW-06060-63 Mfr # 378-2215-OEM
COMPARE
25 PVDF 0.22
Call For Price
Foxx Life Sciences Syringe Filter, 33 mm, 0.22 um PVDF, Sterile; 100/Pk WW-06060-64 Mfr # 378-2415-OEM
COMPARE
33 PVDF 0.22
Call For Price
COMPARE
13 PVDF 0.45
Call For Price
COMPARE
25 PVDF 0.45
Call For Price
Foxx Life Sciences Syringe Filter, 33 mm, 0.45 um PVDF, Sterile; 100/Pk WW-06060-67 Mfr # 378-3415-OEM
COMPARE
33 PVDF 0.45
Call For Price
COMPARE
13 PES 0.22
Call For Price
COMPARE
25 PES 0.22
Call For Price
COMPARE
33 PES 0.22
Call For Price
COMPARE
13 PES 0.45
Call For Price
COMPARE
25 PES 0.45
Call For Price
COMPARE
33 PES 0.45
₹39,400.00INR / Pkg of 100
COMPARE
13 CA 0.22
Call For Price
COMPARE
25 CA 0.22
Call For Price
COMPARE
33 CA 0.22
Call For Price
COMPARE
13 CA 0.45
Call For Price
COMPARE
25 CA 0.45
Call For Price
COMPARE
33 CA 0.45
Call For Price
Showing 1- 18 of 18