This product is now discontinued, View Related Products
Cole-Parmer® Symmetry™ IS Compact Bench Scales
Basic weighing and simple check weighing in a compact design
  • Large backlit display for quick reading
  • Built-in High/Low/OK indicators for fast over/under weighing
  • Display hold function freezes display when load is removed
  • Large stainless steel pan
These affordable compact bench scales handle basic weighing tasks and check weighing operations with a simple keypad and intuitive operation. They offer four weighing units, % weighing, simple parts counting, and check weighing.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
COMPARE
3 6.5 115
Call For Price
COMPARE
6 13 115
Call For Price
COMPARE
15 33 115
Call For Price
COMPARE
30 66 115
Call For Price
COMPARE
3 6.5 230
Call For Price
COMPARE
6 13 230
Call For Price
COMPARE
15 33 230
Call For Price
COMPARE
30 66 230
Call For Price
Showing 1- 8 of 8