Sustayn by Spilfyter Natural Oil-Only Sorbents Rolls
Sorbent material is 100% recycled polypropylene
Go green and support your environmental initiatives with these 100% recycled polypropylene spill rolls. Packaging material is also recycled and printing uses environmentally-friendly water-based inks. Oil-Only sorbents absorb all oil-based liquids and repel water. Ideal for use with lubricants, fuels, mineral oils, and vegetable oils.
32,500.00INR / Bag of 2
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Absorption Length (ft) Width (in)
Availability Pricing
NPS Sustayn by Spilfyter Recycled Oil-Only Natural Heavyweight Perforated Roll, 16" x 150', 2 rolls/Bag WW-86423-15 Mfr # OSR-91
COMPARE
53 gal. (201 L) 150 16
₹32,500.00INR / Bag of 2