Sustayn by Spilfyter Natural Oil-Only Sorbent Pads
Sorbent material is 100% recycled polypropylene
Go green and support your environmental initiatives with these 100% recycled polypropylene spill pads. Packaging material is also recycled and printing uses environmentally-friendly water-based inks. Oil-Only sorbents absorb all oil-based liquids and repel water. Ideal for use with lubricants, fuels, mineral oils, and vegetable oils.
15,700.00INR / Case of 200
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Absorption Description
Availability Pricing
NPS Sustayn by Spilfyter Recycled Oil-Only Natural Lightweight Pad, 16" x 18", 200/Case WW-86423-17 Mfr # OSR-70
COMPARE
29 gal. (110 L) Lightweight pad
₹15,700.00INR / Case of 200