This product is now discontinued, View Related Products
Subminiature Pressure Switches
Low cost and short lead times make this switch perfect for OEM applications
  • Ideal for pneumatic and low pressure hydraulic applications
These compact pressure switches are designed for OEM applications. Made economical with metal blade contacts instead of microswitches, these switches feature Kapton® diaphragms which maintain excellent physical properties over a wide temperature range, as well as offer superb chemical resistance with organic solvents.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
COMPARE
25
Call For Price
COMPARE
60
Call For Price
COMPARE
150
Call For Price
COMPARE
25
Call For Price
COMPARE
60
Call For Price
COMPARE
150