This product is now discontinued, View Related Products
Stuart Tube Rotator
Gentle and effective mixing—ideal for biological suspensions
  • Fully adjustable mixing angle optimizes mixing for any sample
  • Spillage tray keeps workspace clean
  • Tube holders must be purchased separately
These rotators keep cultures in suspension without compromising viability. The angle of rotation is fully adjustable with a scale to measure and reproduce the mixing angle, from horizontal for minimal mixing to vertical for full end-over-end mixing. Wide operating conditions allow use in incubators up to 40°C and in cold rooms down to 4°C.
Variable-speed rotator is best suited to more dynamic laboratory environments with different or changing protocols requiring 2 to 40 rpm. Rotator features a digital speed display and timer providing better repeatability for optimal results. Purchase tube holders separately.
97,900.00 - ₹104,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Tube Rotator with Free Test Tube Holder, 120 VAC
₹104,000.00INR / Each
COMPARE
Tube Rotator with Free Falcon Tube Holder, 120 VAC
₹97,900.00INR / Each