Cole-Parmer® WBS-300 Series Stuart Shaking Water Baths
Customize to your needs with platforms and racks
  • Analog controls and digital temperature display simplify operation
  • Overtemperature and low-level cutoffs improve safety
  • Shaking speed: 20 to 130 rpm
  • Stroke length/orbit size: 3/4" (20 mm)
Use this water bath as a linear or orbital shaker, or a static bath by simply changing the platform. Strong magnets couple to the drive unit so platform changes are quick and easy. The stainless steel shaking platforms with high-tension springs accommodate most bottle and flask sizes. It can hold up to 8 x 250 mL, 6 x 500 mL, or 4 x 1 L Erlenmeyer flasks. For tubes and smaller vessels, order an optional stainless steel rack.
Set speed and temperature using analog dials. Use the LED display to set and view current temperature. A polycarbonate cover (12152-99) or stainless steel cover is required to reach temperatures above 90°C—order separately. Stainless steel tank with integral drain makes cleaning and maintenance fast and efficient.
Choice of platform produces a reciprocating shaking, orbital shaking, or non-shaking system. Order 12152-90, 12152-91, or 12152-92 platform separately.
407,000.00 - ₹411,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (Hz) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
60 120 24
₹411,000.00INR / Each
COMPARE
50 230 24
₹407,000.00INR / Each