Cole-Parmer® DR-200 Series Stuart Rotating Shaker System Drives
Versatile—easily switch out mixing drum for different action
  • Accepts a choice of 5 drums
  • Built in timer
  • Variable speed control 6 to 60 rpm
The rotating shaker system features a variable-speed drive that accepts one of five drums providing different mixing actions to a variety of vessels. Set the analog timer from 10 to 60 minutes or operate the unit continuously.
Required components: 1. Rotator drive, 2. Drum
191,000.00 - ₹216,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Load (lbs) Max Speed (rpm) Power (VAC)
Availability Pricing
COMPARE
6.6 60 120
₹191,000.00INR / Each
COMPARE
6.6 60 230
₹216,000.00INR / Each