Cole-Parmer® SH-200 Series Stuart Microplate Shakers
Provides a fast-mixing action in a tight orbit
  • Nonslip mat and platform lip securely hold plates
  • Built-in timer and digital speed LED display provide reproducible results
  • Can be use in incubators
These microplate shakers combine high speed and a tiny 1.5-mm orbit to produce vigorous shaking perfect for microwell plates. The shakers offer a quit drive mechanism, easy-to-use controller with timer, LED display, and a digital speed selection from 250 to 1250 rpm with a soft start.
Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert. Shaker is capable of continuous use, and the microprocessor maintains speed accuracy over long runs.
Choose the four-plate model for small scale applications or the eight-plate model for high throughput benchtop applications.
93,100.00 - ₹116,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Max Load (lbs) Power (VAC)
Availability Pricing
COMPARE
8 plates 4.4 230
₹116,000.00INR / Each
COMPARE
4 plates 2.2 230
₹93,100.00INR / Each