Cole-Parmer® INC-200 Series Stuart Microplate Incubators
Specially designed for use with microplates and petri dishes
  • Laminar air flow circulation combined with microprocessor control result in excellent temperature uniformity
  • Built-in water tray prevents samples from drying out
  • Up-and-over door for easy access to the chamber
Microplate incubators use forced air circulation to blow preheated air across the shelves for excellent temperature distribution. Quickly set and view the temperature on the bright LED display. Built-in safety feature protects the unit from overheating.
Easily access the interior with the upwards opening acrylic door. The stainless steel chamber is easy to clean and holds four shelves each accommodating 6 plates (or 12 stacked in two layers). Shelves slide out for easy loading. Prevent samples from drying out by adding water to the stainless steel drip tray to increase the humidity within the chamber.
438,000.00 - ₹578,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Max Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
20 80 120
₹578,000.00INR / Each
COMPARE
20 80 230
₹438,000.00INR / Each