Rotor and Tube Adaptors for Cole-Parmer® CF-200 Microcentrifuge
Enhance the performance of your Cole-Parmer® CF-200 Microcentrifuge
The Cole-Parmer CF-200 microcentrifuge includes a fixed-angle rotor that accommodates up to twelve 1.5- or 2.0-mL tubes. Order tube adapters to used 0.2- or 0.5-mL tubes, or order the strip tube rotor to accommodate two 8-place 0.2-mL strip tubes.
18,900.00 - ₹74,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Adapter Shape Type Number Of Tubes Accepted Tube Sizes Accepted
Availability Pricing
COMPARE
Conical 12 0.2 mL
₹18,900.00INR / Each
COMPARE
Conical 12 0.5 mL
₹18,900.00INR / Each
COMPARE
Conical 16 0.2 mL
₹74,800.00INR / Each