Cole-Parmer® HP-200 Series Large-Capacity Digital Hot Plates
Microprocessor controlled, dual thermocouples ensure accurate temperature control
  • Push buttons and dial setting for fast temperature changes and simple control
  • Large plate area accommodates one large vessel or several smaller ones
  • Be safe—hot warning light remains active when plugged in while above 50 °C
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
Microprocessor control allows for accurate plate temperature monitoring. The "Hot" warning light will flash whenever the plate temperature is above 50 °C. An independent safety circuit protects against overheating and internal electronic components are protected against corrosion.
The housing is shaped for stability deflecting spills away from the user. Hot plates also feature BioCote antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.
94,000.00 - ₹284,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
COMPARE
450 230 HP-200D-L-C Digital Hot Plate, Ceramic, 30x30cm, 230 VAC
₹114,000.00INR / Each
COMPARE
450 120 Digital Hot Plate, Ceramic, 12\" X 12\", 120 VAC
₹120,000.00INR / Each
COMPARE
375 230 HP-200D-XL-C Digital Hot Plate, Ceramic, 30x50cm, 230 VAC
₹284,000.00INR / Each
COMPARE
300 230 HP-200D-L-S Digital Hot Plate, Metal, 30x30cm, 230V
₹112,000.00INR / Each
COMPARE
300 120 Digital Hot Plate, Metal, 12\" X 12\", 120V
₹94,000.00INR / Each
COMPARE
300 230 HP-200D-XL-S Digital Hot Plate, Metal, 30x50cm, 230V
₹135,000.00INR / Each
COMPARE
300 120 Digital Hot Plate, Metal, 12\" X 20\", 120V
₹119,000.00INR / Each
Showing 1- 7 of 7